Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cd37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015801.2 | NP_001015801.2 | 7 - 8 | 750 | 87 | |
GenBank mRNA | BC089757.1 | - | 7 - 8 | 750 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |