Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tma7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016374.2 | NP_001016374.1 | 2 - 3 | 83 | 87 | |
GenBank mRNA | BC077652.1 | - | 2 - 3 | 122 | 87 | |
BC135366.1 | - | 2 - 3 | 124 | 87 | ||
CR761261.2 | - | 2 - 3 | 83 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |