Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hdgf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017007.2 | NP_001017007.1 | 2 - 3 | 391 | 115 | |
GenBank mRNA | BC090580.1 | - | 2 - 3 | 391 | 115 | |
CR762281.2 | - | 2 - 3 | 194 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |