Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: polr2d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016248.2 | NP_001016248.1 | 1 - 2 | 81 | 75 | |
XM_018093879.1 | XP_017949368.1 | 7 - 8 | 4920 | 75 | ||
XM_018093880.1 | XP_017949369.1 | 7 - 8 | 4165 | 75 | ||
GenBank mRNA | BC167273.1 | - | 1 - 2 | 371 | 75 | |
BC167274.1 | - | 1 - 2 | 371 | 75 | ||
CR761767.2 | - | 1 - 2 | 81 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |