Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: kti12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016291.2 | NP_001016291.1 | 3 - 4 | 238 | 111 | |
XM_012966570.1 | XP_012822024.1 | 3 - 4 | 432 | 111 | ||
XM_018092083.1 | XP_017947572.1 | 3 - 4 | 271 | 111 | ||
GenBank mRNA | BC170877.1 | - | 2 - 3 | 218 | 111 | |
BC170879.1 | - | 2 - 3 | 218 | 111 | ||
CR761570.2 | - | 3 - 4 | 238 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |