Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: elmod2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016465.2 | NP_001016465.1 | 7 - 8 | 635 | 85 | |
XM_012962363.2 | XP_012817817.1 | 7 - 8 | 725 | 85 | ||
GenBank mRNA | BC166260.1 | - | 7 - 8 | 626 | 85 | |
BC170606.1 | - | 7 - 8 | 635 | 85 | ||
BC170985.1 | - | 7 - 8 | 635 | 85 | ||
CR760937.2 | - | 7 - 8 | 635 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |