Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pgk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016545.2 | NP_001016545.1 | 1 - 2 | 79 | 104 | |
GenBank mRNA | CR760747.2 | - | 1 - 2 | 79 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Xenopus tropicalis Probe spans exons |