Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: crnkl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016895.2 | NP_001016895.1 | 7 - 8 | 1003 | 75 | |
GenBank mRNA | BC155382.1 | - | 6 - 7 | 1176 | 75 | |
CR848544.2 | - | 7 - 8 | 1003 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |