Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ccnq | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012968449.2 | XP_012823903.1 | 2 - 3 | 207 | 93 | |
XM_012968450.2 | XP_012823904.1 | 2 - 3 | 217 | 93 | ||
XM_018096265.1 | XP_017951754.1 | 2 - 3 | 237 | 93 | ||
GenBank mRNA | CR848270.2 | - | 2 - 3 | 294 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Xenopus tropicalis Probe spans exons |