Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: scamp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017190.2 | NP_001017190.1 | 5 - 6 | 506 | 97 | |
GenBank mRNA | CR760611.2 | - | 5 - 6 | 506 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Xenopus tropicalis Probe spans exons |