Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: thrap3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037250.1 | NP_001032327.1 | 12 - 13 | 2742 | 72 | |
XM_012956627.2 | XP_012812081.1 | 13 - 14 | 2868 | 72 | ||
XM_012956628.2 | XP_012812082.1 | 12 - 13 | 2809 | 72 | ||
XM_012956629.2 | XP_012812083.1 | 13 - 14 | 2865 | 72 | ||
XM_018091133.1 | XP_017946622.1 | 13 - 14 | 2939 | 72 | ||
XM_018091134.1 | XP_017946623.1 | 12 - 13 | 2765 | 72 | ||
GenBank mRNA | BC171045.1 | - | 12 - 13 | 2705 | 72 | |
CR759994.2 | - | 12 - 13 | 2742 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |