Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: c19orf24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044450.1 | NP_001037915.1 | 3 - 4 | 415 | 97 | |
XM_012964052.2 | XP_012819506.1 | 3 - 4 | 845 | 97 | ||
GenBank mRNA | CR760950.2 | - | 3 - 4 | 415 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Xenopus tropicalis Probe spans exons |