Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lonrf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045693.1 | NP_001039158.1 | 10 - 11 | 2289 | 80 | |
XM_012957170.2 | XP_012812624.1 | 10 - 11 | 2356 | 80 | ||
GenBank mRNA | BC121307.1 | - | 10 - 11 | 2289 | 80 | |
CR942594.2 | - | 10 - 11 | 2204 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |