Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hnrnpab | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045771.1 | NP_001039236.1 | 2 - 3 | 199 | 66 | |
GenBank mRNA | BC167421.2 | - | 2 - 3 | 215 | 66 | |
CR942727.2 | - | 2 - 3 | 199 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |