Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: eln1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001078709.1 | NP_001072177.1 | 21 - 22 | 1363 | 57 | |
XM_012957185.2 | XP_012812639.1 | 23 - 24 | 1549 | 57 | ||
GenBank mRNA | DQ523561.1 | - | 21 - 22 | 1363 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 Xenopus tropicalis Probe spans exons |