Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sh2d4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079236.2 | NP_001072704.1 | 5 - 6 | 885 | 97 | |
XM_012965810.2 | XP_012821264.1 | 6 - 7 | 1022 | 97 | ||
XM_012965811.2 | XP_012821265.1 | 7 - 8 | 1125 | 97 | ||
XM_012965812.2 | XP_012821266.1 | 6 - 7 | 972 | 97 | ||
XM_012965813.2 | XP_012821267.1 | 5 - 6 | 944 | 97 | ||
GenBank mRNA | BC124048.1 | - | 5 - 6 | 886 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |