Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: esrra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079288.1 | NP_001072756.1 | 6 - 7 | 1272 | 75 | |
XM_012961408.2 | XP_012816862.1 | 6 - 7 | 1008 | 75 | ||
GenBank mRNA | BC125780.1 | - | 6 - 7 | 1272 | 75 | |
BC160530.1 | - | 6 - 7 | 1272 | 75 | ||
CU075398.1 | - | 6 - 7 | 1246 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |