Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: slc18a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079391.1 | NP_001072859.1 | 1 | 1355 | 93 | |
XM_012966764.2 | XP_012822218.1 | 2 | 1747 | 93 | ||
GenBank mRNA | BC123053.1 | - | 1 | 1355 | 93 | |
CU075886.1 | - | 2 | 1619 | 93 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Xenopus tropicalis Both primers and probe map within a single exon |