Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: p3h1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001097231.1 | NP_001090700.1 | 14 - 15 | 2091 | 86 | |
XM_018095197.1 | XP_017950686.1 | 14 - 15 | 2393 | 86 | ||
GenBank mRNA | BC128622.1 | - | 14 - 15 | 2091 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |