Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: prdx5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113054.1 | NP_001106525.1 | 5 - 6 | 456 | 98 | |
GenBank mRNA | BC154896.1 | - | 5 - 6 | 456 | 98 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |