Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ada.2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113897.1 | NP_001107369.1 | 8 - 9 | 893 | 84 | |
XM_012971519.2 | XP_012826973.1 | 8 - 9 | 915 | 84 | ||
GenBank mRNA | BC157229.1 | - | 8 - 9 | 893 | 84 | |
BC171134.1 | - | 7 - 8 | 752 | 84 | ||
BC171136.1 | - | 7 - 8 | 752 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |