Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cables2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114489.1 | NP_001107961.1 | 4 - 5 | 642 | 62 | |
XM_012971718.2 | XP_012827172.1 | 5 - 6 | 765 | 62 | ||
XM_018097721.1 | XP_017953210.1 | 3 - 4 | 599 | 62 | ||
GenBank mRNA | BC088607.1 | - | 4 - 5 | 479 | 62 | |
BC118711.1 | - | 4 - 5 | 619 | 62 | ||
BC159128.1 | - | 4 - 5 | 642 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |