Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: best4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001123442.1 | NP_001116914.1 | 3 - 4 | 662 | 69 | |
GenBank mRNA | BC161756.1 | - | 3 - 4 | 662 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |