Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: grina | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126613.1 | NP_001120085.1 | 5 - 6 | 574 | 69 | |
XM_018094536.1 | XP_017950025.1 | 5 - 6 | 1199 | 69 | ||
GenBank mRNA | BC158526.1 | - | 5 - 6 | 574 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |