Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: clip3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126673.1 | NP_001120145.1 | 2 - 3 | 259 | 60 | |
GenBank mRNA | BC159136.1 | - | 2 - 3 | 259 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |