Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hunk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127077.1 | NP_001120549.1 | 1 - 2 | 733 | 102 | |
GenBank mRNA | BC161479.1 | - | 1 - 2 | 733 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |