Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: slc46a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127138.1 | NP_001120610.1 | 2 - 3 | 1150 | 76 | |
GenBank mRNA | BC161741.1 | - | 2 - 3 | 1150 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |