Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gpatch3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126999.1 | NP_001120471.1 | 4 - 5 | 1086 | 72 | |
GenBank mRNA | BC161307.1 | - | 4 - 5 | 1086 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |