Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: siglec10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127949.1 | NP_001121421.1 | 1233 | 85 | ||
GenBank mRNA | BC166151.1 | - | 1233 | 85 |
Gene Symbol: LOC101732774 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018095750.1 | XP_017951239.1 | 5 - 6 | 716 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |