Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: eln2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142826.2 | NP_001136298.1 | 28 - 29 | 1175 | 67 | |
XM_012970193.2 | XP_012825647.1 | 27 - 28 | 1168 | 67 | ||
XM_012970194.2 | XP_012825648.1 | 27 - 28 | 1153 | 67 | ||
XM_012970195.2 | XP_012825649.1 | 28 - 29 | 1195 | 67 | ||
XM_012970197.2 | XP_012825651.1 | 25 - 26 | 1084 | 67 | ||
GenBank mRNA | CU075851.1 | - | 28 - 29 | 1328 | 67 | |
EU220008.1 | - | 28 - 29 | 1177 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Xenopus tropicalis Probe spans exons |