Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: xyl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111442.1 | NP_001104912.1 | 551 | 91 | ||
GenBank mRNA | AF063865.1 | - | 551 | 91 | ||
AY110911.1 | - | 949 | 91 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Zea mays |