Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hsp82 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001141944.2 | NP_001135416.2 | 155 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Zea mays |