Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gpa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111944.1 | NP_001105414.1 | 47 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Zea mays |