Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NIT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001112112.1 | NP_001105582.1 | 1413 | 66 | ||
GenBank mRNA | AY156979.1 | - | 1413 | 66 | ||
BT064118.1 | - | 1466 | 66 | |||
BT064143.1 | - | 1354 | 66 | |||
BT067700.1 | - | 1382 | 66 | |||
EU966640.1 | - | 1432 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Zea mays |