Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: p5cr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001112317.1 | NP_001105787.1 | 315 | 67 | ||
GenBank mRNA | BT037020.1 | - | 315 | 67 | ||
DQ026301.1 | - | 315 | 67 | |||
EU968742.1 | - | 327 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Zea mays |