Search
Applied Biosystems Diomni Software is an on-premise workflow solution that connects all steps in your real-time PCR (qPCR) research workflows in a single ecosystem.
An intelligent, on-premise platform that connects all the steps of your post-discovery routine qPCR workflows into one single ecosystem, Diomni Software delivers:
In this new era of digitized workflows, Diomni Software powerfully connects the components of your lab's qPCR system in one single, easy-to-use solution.
That's the power of one.
Elevate your digital analytical experience, Streamline biopharma routine qPCR workflows with one connection and one interface with Diomni Software.
Achieve your research goals with the right QuantStudio real-time PCR system for you. Designed for compatibility with TaqMan Assays and application-specific reagents, we make it easy for you to obtain the results you need. Real-time PCR systems
Learn more about the QuantStudio 7 Pro and the QuantStudio 5 real-time PCR systems.
Experience peace of mind in your biopharma lab with our AB Ascend instrument service plan, offering comprehensive scientific support, 24/5 access to priority support, and qualification services to ensure efficiency and compliance in your workflows.
Keep conquering disease. We’ve got your back. We will take extra steps so you can focus on bringing novel therapeutics.
TaqMan Assays are ready-to-use primer-probe pairs that support a wide variety of applications including gene expression, SNP genotyping, and microbial detection to meet your research needs. Our portfolio includes over 20 million predesigned assays, each covered by our TaqMan Guarantee, with options to customize your own assay.
Applied Biosystems master mixes and reagents are backed by over 25 years of expertise and innovation of real-time PCR reagents.
Our master mixes deliver exceptional sensitivity and accuracy in a wide variety of research applications including but not limited to gene expression analysis, microRNA analysis, and virus detection.
Learn more about qPCR master mixes and reagents for research workflows ›
Order TaqMan master mixes and SYBR Green master mixes labeled “For Research Use Only”.
Diomni Software offers advanced workflow functionality.
Instrument integration |
|
Integration |
|
Workflow support |
|
Interpretive results |
|
| Data management |
|
Compatibility | QuantStudio 7 Pro Real-Time PCR System QuantStudio 5 Real-Time PCR System |
Block type | 96-well 384-well Taqman Array Card (TAC) |
Operating system | Microsoft Windows™ 11 or Microsoft Windows 10 operating system for Diomni Software |
Supported browsers | Google Chrome™ Microsoft Edge™ Apple Safari™ (OS X) Mozilla Firefox™ |
Security and compliance | SAE Administrator Console v3 |
Supported workflows | RUO (Research Use Only) |
For Research Use Only. Not for use in diagnostic procedures.