Search
Announcing EVOS M3000 Software Revision 1.0.0.57458
Revision 1.0.0.57458 provides the following changes for existing EVOS M3000 users.
Please review the Installation Instructions before updating your software.
Changes
The software is provided preinstalled on the new Invitrogen EVOS M3000 Imaging System.
The software upgrade to the EVOS M3000 imaging system can be executed either automatically or manually. Both methods are outlined below.
IMPORTANT! It is not possible to downgrade the instrument software.
NOTE: The upgrade process will take approximately 5 minutes to complete.
NOTE: All user defined settings will stay the same and will not be modified by the upgrade.
In case there is an issue encountered during the upgrade process, try again. If the issue persists, contact service. For service information, refer to the contact information found from the EVOS M3000 Imaging System User Guide (Publication Number MAN0030066).
Automatic Software Upgrade
Automatic software upgrade is available if the instrument is connected to the Thermo Fisher Connect Platform. Refer to the EVOS M3000 Imaging System User Guide for instructions about creating a connection to Thermo Fisher Connect Platform.
When the EVOS M3000 Imaging System has been connected to the Connect software, upgrade can be carried out with the following steps.
IMPORTANT! Remove all external equipment from the USB ports before you install the software update.
IMPORTANT! Do not switch the instrument off during the software installation. The installation time depends on the software update. During the installation, the instrument may re-boot several times. The installation is complete when the Capture screen re-opens.
a. Prompt opens asking if you want to start the upgrade. Select Yes. The upgrade starts automatically
b. Wait for the upgrade to complete. The upgrade takes approximately 5 minutes in addition to the time needed to download the software upgrade file. The download time depends on the connection speed.
NOTE: Do not interrupt the process.
c. The system restarts when the upgrade is ready.
Manual Software Upgrade
If there is no connection to Thermo Fisher Connect Platform available, EVOS M3000 can be upgraded to v1.0.0.57458 manually with the following steps.
IMPORTANT! Remove all external equipment from the USB ports before you install the software update.
IMPORTANT! Do not switch the instrument off during the software installation. The installation time depends on the software update. During the installation, the instrument may re-boot several times. The installation is complete when the Capture screen re-opens.
a. There will be a prompt asking if you want to start upgrade, select Yes. Upgrade should start automatically.
b. Wait for the upgrade to be completed. The upgrade takes approximately 5 minutes
NOTE: Do not interrupt the process.
c. The system restarts when the upgrade is ready.
IMPORTANT! If you experience any issues with the installation or find that it is not completing as expected, please reboot your instrument and contact Technical Support.
The number for your local Technical Support Team can be found by navigating to our home page.
For Research Use Only. Not for use in diagnostic procedures.