Search
The Pathatrix™ Auto System has obtained AFNOR™ certification for the ‘Pathatrix™ Auto Salmonella spp.’ Validation data demonstrates equivalence to the reference method ISO 6579 (2002).
Food categories:
Meat products: including raw beef, ready-to-eat and ready-to-reheat meat products
Dairy products: including heat-treated milk and UHT, canned or pasteurized dairy products
Salmonella detection workflows:
Two protocols have been validated:
Both protocols allow the screening of the negative samples typically within 2 days, while 3 days are required for the positive samples; compared to 4 days for culture negatives following ISO.
The Pathatrix Auto Instrument is a recirculation immunomagnetic separation system. Using magnetic beads coated with antibodies, the Pathatrix Auto Instrument quickly concentrates the target organisms such as Salmonella from the enriched samples, and the on-line wash step removes non-target organisms and any PCR inhibitors. This helps reduce the time to results and enables increased pathogen testing at lower costs.
Applying the Pathatrix Auto System full workflows in your food testing lab enables:
For testing of Food and Environmental samples only
©2013 Life Technologies Corporation. All rights reserved. The trademarks mentioned herin are the property of Life Technologies Corporation and/or their respective owners.