Endpoint PCR (PCR) is a method used to determine if a gene of interest is expressed in a sample. Once cDNA is made through reverse transcription, it is amplified using a DNA polymerase-based reaction in a thermal cycler. Quality enzymes and precise temperature control improve the specificity and reproducibility of results. Select from a broad range of enzymes, for a solution to every challenge; and industry-renowned thermal cyclers that meet your lab needs. Thermo Fisher Scientific also offers a comprehensive portfolio of nucleic acid electrophoresis products, whether you prefer pre-cast or pour-you-own gels.
Thermal cyclers
Our engineers have been designing and manufacturing high-quality thermal cyclers since 1987, earning a reputation for reliability, accuracy, and user-friendly interfaces. We offer a thermal cycler for every challenge, application, and budget.
ProFlex PCR System
NEWVeritiPro
Thermal Cycler
SimpliAmp Thermal Cycler
MiniAmp Thermal Cycler
Automated Thermal Cycler (ATC)
ProFlex PCR System
VeritiPro Thermal Cycler
SimpliAmp Thermal Cycler
MiniAmp Thermal Cyclers
Automated Thermal Cycler
Block formats
3 x 32-well 0.2 mL
96-well 0.2 mL
2 x 96-well 0.2 mL
2 x flat block
2 x 384-well 0.02 mL
96-well 0.2 mL
96-well 0.2 mL
96-well 0.2 mL
96-well 0.2 mL
384-well 0.02 mL
Tubes, plates & accessories
Small-scale experiments with a few samples
Daily experiment needs
Complete workflow experiment needs, ideal for automation
Single tubes, strips, caps, adhesive film & accessories
MicroAmp optical microplates
MicroAmp EnduraPlate optical microplates
  • Single tubes
  • Single tubes with caps
  • 8-strip tubes with separate caps
  • 8-strip tubes with attached caps
  • 12-strip caps
  • 32-well
  • 48-well Fast
  • 96-well
  • 96-well Fast
  • 384-well
  • 96-well
  • 96-well Fast
  • 384-well
DNA polymerases
For gene expression assays, use hot-start PCR enzymes such as Invitrogen Platinum II Taq and Applied Biosystems AmpliTaq Gold DNA polymerases, both designed for enhanced target specificity.
Platinum II Taq
Hot-Start DNA Polymerase
AmpliTaq Gold 360 DNA Polymerase
Hot-start technology
AntibodyChemical
Enzyme activation time
2 min10 min
Universal annealing temperature of 60°C
YesNo
DNA synthesis speed
15 sec/kb60 sec/kb
Flexible extension step*
YesNo
Inhibitor tolerance
YesNo
Amplification length
Up to 5 kbUp to 5 kb
GC-rich format
YesYes
Stand-alone enzyme
ColorlessColorless
Master mix format
Colorless Green**Colorless
* The extension step can be extended up to 60 sec/kb without the effect of specificity.
** Contains electrophoresis tracking dyes and a density reagent for direct gel loading of PCR products.
† Green buffer available as separate item for use with stand-alone enzyme.
Nucleic acid electrophoresis
Instrumentation
Electrophoresis gels
Stains
Molecular weight markers
Learn about Invitrogen nucleic acid electrophoresis instruments for low to high throughput; and imagers to capture results.
Choose from pre-set E-gel cassettes, with a range of throughput and stain options, run on the E-Gel PowerSnap Device; or pour-your-own gel powders and reagents. 
Find highly sensitive DNA staining reagents with formats include: SYBR Gold, SYBR Green I, SYBR Green II, and SYBR Safe dyes.
Choose your ladders from a wide variety of size ranges, applications, and formats – all exceeding industry standards.
Style Sheet for Global Design System
Style Sheet for Komodo Tabs