Search
Thermo Scientific Lab Freezers facilitate reliable, secure storage for your sample storage needs in a wide variety of application-specific configurations.
Select from the categories below, based on the type of samples you’re storing, then choose the from specific models meet your performance and feature configuration needs.
Auto defrost (-30°C) |
Manual defrost (-20°C) |
Blood/plasma |
Flammable materials |
Vaccine storage |
Description |
||||
|---|---|---|---|---|
Routine to complex sample storage in demanding, and space-challenged labs. Automatic defrost eliminates need for periodic sample removal. |
Routine to complex storage where samples cannot tolerate even slight cabinet temperature fluctuations that may occur during auto-defrost cycles. | AABB, ANRC AND FDA compliant for Typicstorage of whole blood, blood components, and plasma samples. | NFPA and OSHA compliant safe storage for volatiles, compounds, and solvents when a spark-free environment is required. | High performance perseveration of vaccine efficacy and potency by adhering to strict cold chain requirements and vaccine specific tolerances. |
Typical applications |
||||
| Reagents, daily cell culture media, staining solutions, membranes. | Enzymes, pharmaceuticals, and samples requiring -20°C storage. | Whole blood, blood components, plasma. | Volatiles, flammable or explosive compounds, and solvents requiring a spark-free environment. | Vaccines |
Capacity |
||||
11.5 - 29.2 cu. ft. 326 - 827L |
5 - 29.2 cu. ft. 141.5 - 827L |
4.9-51.1 cu. ft. 133L - 1447L |
5 - 23 cu. ft. 141.6L- 651L |
1.1-51.1 cu. ft. 32L - 1447L |
Size configurations |
||||
Undercounter
Singe door upright
Double door upright |
Countertop
Undercounter
Singe door upright
Double door upright |
Undercounter
Singe door upright |
Undercounter
Singe door upright
Double door upright |
Countertop
Undercounter
Singe door upright
Double door upright |