Search
The initial generation of the primary genetic sequence of a particular organism is called de novo sequencing. A detailed genetic analysis of any organism is possible only after de novo sequencing has been performed.
De novo sequencing is typically accomplished by assembling individual sequence reads into longer contiguous sequences (contigs) or correctly ordered contigs (scaffolds) in the absence of a reference sequence.
Historically, de novo sequencing was carried out using capillary electrophoresis (CE) sequencers. With its long read lengths and high accuracy, CE-based sequencing made overlap consensus assembly the gold-standard technology for de novo projects. However, more recently, the high-throughput capabilities of massively parallel sequencing and the development of short-read assemblers have significantly reduced the time and cost associated with sequencing an entire genome.
De novo sequencing yields a primary genetic sequence of a particular organism where it may not previously be available. It is necessary to first perform de novo sequencing in order to further analyze the genetic information of an organism in more detail. As the sequencing of an organism can be a lengthy and costly endeavor through traditional sequencing methods, the capabilities of next generation sequencing technologies empower the progress of de novo sequencing efforts.