Search
SYBR GreenER qPCR SuperMix Universal is a ready-to-use cocktail containing all components, except primers and template, for the amplification and detection of DNA in real-time quantitative PCR (qPCR). It combines a chemically modified “hot-start” version of Taq DNA polymerase with integrated uracil DNA glycosylase (UDG) carryover prevention technology and a novel fluorescent dye to deliver excellent sensitivity in the quantification of target sequences, with a linear dose response over a wide range of target concentrations. Volumes are provided for 100 or 500 amplification reactions of 50 μl each. SYBR GreenER qPCR SuperMix Universal is supplied at a 2X concentration and contains hot-start Taq DNA polymerase, SYBR GreenER fluorescent dye, MgCl2, dNTPs (with dUTP instead of dTTP), UDG, and stabilizers. The SuperMix formulation can quantify fewer than 10 copies of a target gene, has a broad dynamic range, and is compatible with melting curve analysis.
| Component | 100-rxn Kit | 500-rxn Kit |
| SYBR GreenER qPCR SuperMix Universal | 2 × 1.25 ml | 12.5 ml |
| ROX Reference Dye | 100 μl | 500 μl |
Kit components are shipped on dry ice and should be stored at 4ºC. Storage at –20ºC may extend shelf life.
Minimize exposure of both the SuperMix and ROX Reference Dye to direct light. Exposure to direct light for an extended period of time may result in loss of fluorescent signal intensity.
This SuperMix is also included with the SYBR GreenER Two-Step qRT-PCR Kit Universal, catalog nos. 11765-100 and 11765-500.
This kit can be used with a variety of real-time instruments, including but not limited to the ABI PRISM 7000, 7700, and 7900HT; the ABI 7300 and 7500 Real-Time PCR Systems; the ABI GeneAmp 5700; the Bio-Rad iCycler the Stratagene Mx3000P, Mx3005P, and Mx4000 the Corbett Research Rotor-Gene the MJ Research DNA Engine Opticon, Opticon 2, and Chromo 4 Real-Time Detector; and the Cepheid Smart Cycler. Optimal cycling conditions will vary with different instruments.
Use up to 100 ng of genomic DNA or 10–107 copies of plasmid DNA in a 10-μl volume. Note that 1 μg of plasmid DNA contains 9.1 × 1011 copies divided by the plasmid size in kilobases.
Primer design is one of the most important parameters when using SYBR GreenER qPCR SuperMix. We strongly recommend using a primer design program such as OligoPerfect, available here, or Vector NTI. When designing primers, the amplicon length should be approximately 80–250 bp. A final concentration of 200 nM per primer is effective for most reactions. Optimal results may require a titration of primer concentrations between 100 and 500 nM.
The hot-start DNA polymerase is activated in the 10-minute incubation at 95°C before PCR cycling. Melting Curve Analysis Melting curve analysis should always be performed following real-time qPCR to identify the presence of primer dimers and analyze the specificity of the reaction. Program your instrument for melting curve analysis using the instructions provided with your specific instrument.
Magnesium chloride is included in the SuperMix at an optimized concentration for qPCR.
ROX Reference Dye can be included in the reaction to normalize the fluorescent reporter signal for instruments that are compatible with that option. ROX is supplied at a 25 μM concentration, and is composed of a glycine conjugate of 5-carboxy-X-rhodamine, succinimidyl ester in 20 mM Tris-HCl (pH 8.4), 0.1 mM EDTA, and 0.01% Tween 20. Use the following table to determine the amount of ROX to use with a particular instrument:
| Instrument | Amount of ROX per 50-μl reaction | Final ROX Concentration |
| ABI 7000, 7300 7700, and 7900HT | 1.0 μl | 500 nM |
| ABI 7500; Stratagene Mx3000, Mx3005P, and Mx4000 | 0.1 μl* | 50 nM |
* To accurately pipet 0.1 μl per reaction, we recommend that you dilute ROX 1:10 immediately before use and use 1 μl of the dilution.
Note: SYBR GreenER qPCR SuperMix for ABI PRISM includes ROX in the SuperMix at a 500 nM final concentration (see Additional Products in ordering table)
The Bio-Rad iCycler requires the use of fluorescein as a reference dye to normalize the fluorescent reporter signal in reactions with SYBR GreenER qPCR SuperMix. Fluorescein NIST-Traceable Standard is available separately as a 50-μM solution (see Additional Products, page 1). We recommend using a final concentration of 50 nM as a general starting point in qPCR. Optimal results may require a titration between 10 and 100 nM. SYBR GreenER qPCR SuperMix for iCycler includes fluorescein in the SuperMix at an optimized concentration for the iCycler (see Additional Products in ordering table)
Follow the general protocol below for qPCR on ABI real-time instruments. Note the lower amount of ROX Reference Dye required for the ABI 7500. This generic protocol may also be used as a starting point for other real-time instruments.
- 50ºC for 2 minutes hold (UDG incubation)
- 95ºC for 10 minutes hold (UDG inactivation and DNA polymerase activation)
- 40 cycles of:
- 95ºC, 15 seconds
- 60ºC, 60 seconds
- Melting curve analysis: Refer to instrument documentation
| Component | Single rxn | Notes |
| SYBR GreenER qPCR SuperMix Universal | 25 μl | 1X final conc. |
| Forward primer, 10 μM | 1 μl | 200 nM final conc. |
| Reverse primer, 10 μM | 1 μl | 200 nM final conc. |
| ROX Reference Dye (optional) | 1 μl/0.1 μl | See table above |
| Template (up to 100 ng of genomic DNA, 10–107 copies of plasmid DNA, or cDNA generated from up to 1 μg of total RNA) | 5–10 μl | 5–10 μl |
| DEPC-treated water | DEPC-treated water |
| Problem | Possible Cause | Solution |
|---|---|---|
| Signals are present in no-template controls, and/or multiple peaks are present in the melting curve graph | Template or reagents are contaminated by nucleic acids (DNA, cDNA) | Use melting curve analysis and/or run the PCR products on a 4% agarose gel after the reaction to identify contaminants. Take standard precautions to avoid contamination when preparing your PCR reactions. Ideally, amplification reactions should be assembled in a DNA-free environment. We recommend using aerosol-resistant barrier tips. |
| Primer dimers or other primer artifacts are present | Use melting curve analysis to identify primer dimers. We recommend using validated pre-designed primer sets or design primers using dedicated software programs or primer databases. Primer contamination or truncated or degraded primers can lead to artifacts. Check the purity of your primers by gel electrophoresis | |
| No amplification curve appears on the qPCR graph | There is no PCR product | Run the reaction on a gel to determine whether PCR worked. Then proceed to the troubleshooting steps below. |
| No PCR product is evident, either in the qPCR graph or on a gel | The protocol was not followed correctly | Verify that all steps have been followed and the correct reagents, dilutions, volumes, and cycling parameters have been used. |
| Template contains inhibitors, nucleases, or proteases, or has otherwise been degraded. | Purify or re-purify your template. | |
| Primer design is suboptimal | Verify your primer selection. We recommend using validated pre-designed primers or design primers using dedicated software programs or primer databases. | |
| PCR product is evident in the gel, but not on the qPCR graph | qPCR instrument settings are incorrect | Confirm that you are using the correct instrument settings (dye selection and calibration, reference dye, filters, acquisition points, etc.). |
| Problems with your specific qPCR instrument | See your instrument manual for tips and troubleshooting. | |
| PCR efficiency is above 110% | Template contains inhibitors, nucleases, or proteases, or has otherwise been degraded. | Purify or re-purify your template. Inhibitors in the template may result in changes in PCR efficiency between dilutions |
| Nonspecific products may be amplified. | Use melting curve analysis if possible, and/or run the PCR products on a 4% agarose gel after the reaction to identify contaminants. Suboptimal primer design may lead to nonspecific products. Use validated pre-designed primers or design primers using dedicated software programs or primer databases. | |
| PCR efficiency is below 90% | The PCR conditions are suboptimal | Verify that the reagents you are using have not been freeze-thawed multiple times and have not remained at room temperature for too long. Verify that the amount of primers you are using is correct. |