Technical Resources - Media Formulations
14180 - HBSS, 10x, no calcium, no magnesium, phenol red
  • Catalog Number(s)
ComponentsMolecular WeightConcentration (mg/L)mM
Inorganic Salts
Potassium Chloride (KCl)75.04000.053.333332
Potassium Phosphate monobasic (KH2PO4)136.0600.04.4117646
Sodium Chloride (NaCl)58.080000.01379.3103
Sodium Phosphate dibasic (Na2HPO4-7H2O)268.0900.03.358209
Other Components
D-Glucose (Dextrose)180.010000.055.555557
Phenol Red398.0100.00.2512563
Reference:
1. Hanks, J.H. and Wallace, R.E. (1949) Relation of Oxygen and Temperature in the Preservation of Tissues by Refrigeration. Proc. Soc. Exp. Biol. Med., 71:196.
Related Products:
14170 - HBSS, no calcium, no magnesium
Cat No: 14170070 , 14170088 , 14170112 , 14170120 , 14170138 , 14170146 , 14170161
14175 - HBSS, no calcium, no magnesium, no phenol red
Cat No: 14175046 , 14175053 , 14175079 , 14175095 , 14175103 , 14175129 , 14175137 , 14175145
14185 - HBSS, 10x, no calcium, no magnesium, no phenol red
Cat No: 14185045 , 14185052