CD309 (FLK1) Antibody (17582181) in Flow
Staining of the bEnd-3 cell line with 0-25 ug of Rat IgG2a K Isotype Control APC (Product # 17-4321) (open histogram) or 0-25 ug of Anti-Mouse CD309 (FLK1) APC (filled histogram). Total viable cells were used for analysis.
Staining of the bEnd-3 cell line with 0-25 ug of Rat IgG2a K Isotype Control APC (Product # 17-4321) (open histogram) or 0-25 ug of Anti-Mouse CD309 (FLK1) APC (filled histogram). Total viable cells were used for analysis.
Related Products
Related Images