Search
Explore the wide range of Multidrop Reagent Dispensers, designed to meet the diverse needs of your laboratory workflows.
Multidrop Combi SMART+ |
|
|
|
|
| User interface |
||||
On-board interface |
On-board interface |
On-board interface |
PicoIT PC software |
PicoIT PC software |
| Dispensing Volume | ||||
0.5-2500 µL |
0.5-2500 µL |
50 nL-50 µl |
8 pL - 10 µL |
11 pL - 10 µL |
| Plate format | ||||
6- to 1,536-well plates |
6- to 1,536-well plates |
6- to 1,536-well plates |
12, 24, 48, 96, 384, 1536 well plate
|
12, 24, 48, 96, 384, 1536 well plate
|
| Plate height adjustment | ||||
Automatic |
Automatic |
Automatic |
Automatic |
Automatic |
| Automation/robot compatible | ||||
Yes |
Yes |
Yes |
Yes |
No |
| Dimensions | ||||
| 14.0 x 8.6 x 12.9 in. (35.6 x 21.8 x 32.8 cm) |
14.0 x 8.6 x 12.9 in. (35.6 x 21.8 x 32.8 cm) |
14 x 8.6 x 14.8 in.
|
18.5 x 15.0 x 9.1 in. (47 x 38 x 23 cm) | 15.0 x 11.0 x 7.5 in. (38.5 x 28 x 19 cm) |
| Weight | ||||
20.1 lbs/9.1 kg
|
20.1 lbs/9.1 kg
|
21.2 lbs/9.6 kg |
31.0 lbs./14.1 kg |
13.0 lbs/5.9 kg |
*Multidrop Combi SMART+ uses RFID to track cassette usage
For Laboratory Use. It is the customer’s responsibility to ensure that the performance of the product is suitable for customers’ specific uses or applications.