Search
HeLa cells were seeded in Nunc™ Microwell 96-well optical bottom plates (Cat# 164588) and co-transduced with CellLight® ER-GFP and CellLight® Golgi-RFP reagents. After incubation overnight, cells were imaged on an EVOS® FL Auto imaging system with a 40X coverslip-corrected objective.