Choose the right Thermo Scientific belt scale system for you
Still have questions? We're here to help!
Contact a sales representative
  Ramsey IDEA
Belt Scale System
Ramsey Series 20
Belt Scale System
Ramsey Series 17
Belt Scale System
Ramsey Series 14
Belt Scale System
Accuracy +/-1% +/-0.5% +/-0.25% +/-0.125%
Number of idlers 1 1 or 2 2 or 4 3, 4, or 6
Number of load cells 1 or 2 1 or 2 2 4
Belt widths 18"–71" 16"–78" 18"–102" 18"–102"
HD version available for larger sizes
Belt speed range:
Material must spend at least 0.5 seconds in the weigh area; varies dending on # of idlers and idler spacing.
Belt loading
Model 10-101-R1: 16.5–250 lbs
Model 10-101-R2: 30–750 lbs
Model 10-20 1:  50–1800 lbs
Model 10-20-CWT: 20–50 lbs
Contact a rep for calculation
8–750 lbs/ft
Idler spacing
R1 - Larger than 21.5"
R2 - Larger than 22.5"
32" minimum
Standards 36", 42", 48", 900mm, 1000mm and 1200mm
Standards 36", 42", 48", 900mm, 1000mm and 1200mm
Types of applications Cost-effective weighing for non-critical or lower value materials, modular installation General in plant or in-process High-accuracy weighing
High-value materials
High quality and high accuracy requirement
Certifiable or basis-of-payment 
If your application doesn't fit one of the above scales, please contact our factory to discuss a custom application.