Protein Digest
Making protein sample preparation easy
Biomarker and biotherapeutic characterization and quantitation just got a lot easier. Use the Thermo Scientific™ SMART Digest™ and SMART Digest Immunoaffinity (IA) kits for protein sample preparation that is fast, simple, highly reproducible, and sensitive.
All kits provide:
- Fast, easy, highly reproducible, characterization, and quantitation of proteins
- In a range of scalable kit options to meet your workflow requirements.
| SMART Digest Trypsin Kits | Uses trypsin to cleave the C-terminal side of lysine and arginine amino acid residues of peptides. |
| SMART Digest Chymotrypsin Kits | Uses chymotrypsin to cleave the peptide bonds formed by aromatic residues such as tyrosine, phenylalanine, and tryptophan. |
| SMART Digest Proteinase K Kits | Uses proteinase K kits to cleave the peptide bond adjacent to the carboxyl group of aliphatic and aromatic amino acids with blocked alpha amino groups. |
| SMART Digest ImmunoAffinity (IA) Streptavidin Kits | The Streptavadin version of the kit allows for purification of various biomolecules via the formation of strong streptavidin-biotin bond. |
| SMART Digest ImmunoAffinity (IA) Protein A Kits | The Protein A version of the kit allows for targeted purification of immunoglobulins due to the strong affinity for binding to the Fc portion of the molecule. |
| SMART Digest ImmunoAffinity (IA) Protein G Kits | The Protein G version of the kit allows for targeted purification of immunoglobulins due to the strong affinity for binding to the Fab and Fc portion of the molecule. |
SMART Digest kit:
- Is significantly faster
- Is reproducible protein digestion
- Provides improved sequence coverage
- Provides improved data quality
- Significantly increased speed of sample processing compared to in-solution digestion
- 3-step sample preparation protocol
- Higher throughput
- Ability to automate
- More reproducible than in-solution digestion
- Higher sensitivity
Fast and reproducible protein digestion for improved sequence coverage and data quality over traditional in-solution digestion. This quick introductory video reviews:
- The easy protocol
- Why it’s simpler, quicker, and reproducible—producing quality data
- Its application in biopharmaceutical and protein research
Learn how SMART Digest kits provide easy, fast, and highly reproducible protein digestion compared to in-solution digestion protocols with this introduction animation.
- 3-step sample preparation protocol
- Higher throughput
- Ability to automate
- Higher sensitivity