When a DNA sequence is presented in scientific journals and databases, it is usually written as a single strand. By convention, the strand shown is the coding (+) or sense strand, identical in sequence (with T's instead of U's) to its mRNA copy. The mRNA then serves as a template for translation, its 5' or upstream sequence (beginning with AUG) corresponding to the NH3-terminal "Met" of the protein.
schematic of an example of a DNA strand being transcribed into mRNA, which would then be translated to protein
The complementary DNA strand is called the noncoding or (-) strand and corresponds to the antisense strand. In order to hybridize with and thus detect a specific mRNA, the probe must consist of complementary antisense sequence. Antisense RNA probes are generated when an RNA phage promoter adjacent to more downstream sequence is used.
For Research Use Only. Not for use in diagnostic procedures.