Search
| Organism Type | Human | The WiT49 cell line was derived from a primary lung metastasis of an aggressive Wilms tumor.2 WiT49 cells were maintained in 1:1 high-glucose Dulbecco’s modified Eagle’s medium/ nutrient mixture F-12, 10% fetal calf serum, 100 units/ml penicillin, and 100 mg/ml streptomycin. Transfections were done using LipofectAMINE Plus . Stably transfected cells were selected with 1 mg/ml G418 (Life Technologies, Inc.) and maintained in 0.6 mg/ml G418 |
| Tissues | Lung | |
| Phenotype | Either | |
| Primary | yes | |
| Application | Cell Culture / Growth Conditions,Transfection: Gene Expression |
Maintaining healthy cells is the key to experimental success and reproducible research results. To give you confidence in the health of your cells every step of the way, we’ve highlighted the technologies and products within cell biology that are critical to maintaining optimal cell health. No matter how you are using your cells, you can count on these products to help keep them healthy.
|
|
|
| |||